2,5-Dimethyl-1,3,4-thiadiazol

Lieferant: Thermo Scientific

JFD03118.250 JFD03118.1
MAYBJFD03118.250EA 0 EUR
MAYBJFD03118.250 MAYBJFD03118.1
2,5-Dimethyl-1,3,4-thiadiazol
2,5-Dimethyl-1,3,4-thiadiazole
Formel: C₄H₆N₂S
Molecular Weight: 114,17 g/mol
MDL: MFCD00020722
CAS-Nummer: 27464-82-0
EINECS: 248-473-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR