2,2'-Bithiophen

Lieferant: FLUOROCHEM

Synonyme: 2,2′-Dithienyl, 2,2′-Bithienyl

001054-25G 001054-5G
FLUO001054-25GEA 112 EUR
FLUO001054-25G FLUO001054-5G
2,2'-Bithiophen
2,2'-Bithiophen
Formel: C₈H₆S₂
Molecular Weight: 166,27 g/mol
MDL: MFCD00005414
CAS-Nummer: 492-97-7
EINECS: 207-767-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR