2,2':5',2''-Terthiophen ≥99%

Lieferant: Thermo Scientific

A16748.03 A16748.06 A16748.MD
A16748.03EA 225 EUR
A16748.03 A16748.06 A16748.MD
2,2':5',2''-Terthiophen ≥99%
2,2':5',2''-Terthiophen

Purity: 99% CAS No.: 1081-34-1 Molecular Formula: C12H8S3 Formula Weight: 248.39 Melting Point: 89-94° Boiling Point: 160°/0.1mm Merck Reference: 14,9174 MDL No.: MFCD00012167

Formel: C₁₂H₈S₃
Molecular Weight: 248,39 g/mol
Siedepunkt: 160 °C (0,1 mmHg)
Schmelzpunkt: 89…94 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012167
CAS-Nummer: 1081-34-1
Merck Index: 14,9174

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR