3-(Trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Trifluormethyl)benzolboronsäure, Dihydroxy(3-(trifluormethyl)phenyl)boran, (3-(Trifluormethyl)phenyl)metaborsäure

BB-2378-25G BB-2378-100G BB-2378-5G
COBBBB-2378-25GEA 121 EUR
COBBBB-2378-25G COBBBB-2378-100G COBBBB-2378-5G
3-(Trifluormethyl)phenylboronsäure
3-(Trifluormethyl)phenylboronsäure
Formel: C₇H₆BF₃O₂
Molecular Weight: 189,93 g/mol
MDL: MFCD00151854
CAS-Nummer: 1423-26-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR