(3,4-Dichlorphenyl)essigsäure

Lieferant: Thermo Scientific

Synonyme: 3,4-Dichlorophenylacetic acid

SB01855.1 SB01855.10 SB01855.50
MAYBSB01855.1EA 0 EUR
MAYBSB01855.1 MAYBSB01855.10 MAYBSB01855.50
(3,4-Dichlorphenyl)essigsäure
(3,4-Dichlorphenyl)essigsäure
Formel: (C₆H₃Cl₂)CH₂COOH
Molecular Weight: 205.04 g/mol
Siedepunkt: 314
Schmelzpunkt: 91…92 °C
MDL: MFCD00004333
CAS-Nummer: 5807-30-7
EINECS: 227-368-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR