3-Ethinylanisol 96%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
1-Methoxy-3-ethinylbenzol, 1-Ethinyl-3-methoxybenzol, (3-Methoxyphenyl)acetylen, (3-Methoxyphenyl)ethin
H55254.MD
H55254.03
H55254.06
H55254.MDEA
29.1
EUR
H55254.MD
H55254.03
H55254.06
3-Ethinylanisol 96%
3-Ethinylanisol
Purity: 96%
CAS No.: 768-70-7
Molecular Formula: C9H8O
Formula Weight: 132.16
Storage Temperature: 2°C to 8°C
Flash Point: 76°(169°F)
Refractive Index: 1.5550
Boiling Point: 204-210°
MDL No.: MFCD00160810
Formel:
C₉H₈O Molecular Weight: 132,16 g/mol Siedepunkt: 204…210 °C Dichte: 1,04 Flammpunkt: 76 °C (169 °F) |
MDL:
MFCD00160810 CAS-Nummer: 768-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...