3-Fluor-2-pyridincarbonitril

Lieferant: COMBI-BLOCKS

Synonyme: 2-Cyan-3-fluorpyridin, 3-Fluor-2-pyridincarbonsäurenitril

OS-7581-5G OS-7581-25G OS-7581-1G
COBBOS-7581-5GEA 63 EUR
COBBOS-7581-5G COBBOS-7581-25G COBBOS-7581-1G
3-Fluor-2-pyridincarbonitril
3-Fluor-2-pyridincarbonitril
Formel: C₆H₃FN₂
Molecular Weight: 122,1 g/mol
MDL: MFCD06797501
CAS-Nummer: 97509-75-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR