2-Indolcarbaldehyd ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Indol-2-carboxaldehyd
H54638.03
H54638.06
H54638.03EA
109
EUR
H54638.03
H54638.06
2-Indolcarbaldehyd ≥97%
2-Indolcarbaldehyd
Purity: 97%
CAS No.: 19005-93-7
Molecular Formula: C9H7NO
Formula Weight: 145.16
Melting Point: 138-142°
MDL No.: MFCD03001425
Formel:
C₉H₇NO Molecular Weight: 145,16 g/mol Schmelzpunkt: 138…142 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03001425 CAS-Nummer: 19005-93-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...