6-Propyl-2-thioxo-2,3-dihydro-4(1H)-pyrimidinon

Lieferant: Thermo Scientific
Achtung

Synonyme: Propylthiouracil, 4-Hydroxy-2-mercapto-6-propylpyrimidin, 4-Propyl-2-thiouracil

SB00664.1 SB00664.10 SB00664EE
MAYBSB00664.1EA 0 EUR
MAYBSB00664.1 MAYBSB00664.10 MAYBSB00664EE
6-Propyl-2-thioxo-2,3-dihydro-4(1H)-pyrimidinon
6-Propyl-2-thiouracil
Formel: C₇H₁₀N₂OS
Molecular Weight: 170.24 g/mol
Siedepunkt: 448
Schmelzpunkt: 215…218 °C
MDL: MFCD00006041
CAS-Nummer: 51-52-5
EINECS: 200-103-2
Merck Index: 12,08054

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR