2-(Brommethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Brommethyl)phenyl)metaborsäure, Dihydroxy(2-(brommethyl)phenyl)boran, 2-(Brommethyl)benzolboronsäure

BB-2467-5G BB-2467-25G BB-2467-1G
COBBBB-2467-5GEA 0 EUR
COBBBB-2467-5G COBBBB-2467-25G COBBBB-2467-1G
2-(Brommethyl)phenylboronsäure
2-(Brommethyl)phenylboronsäure
Formel: C₇H₈BBrO₂
MDL: MFCD01318941
CAS-Nummer: 91983-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR