4-(Brommethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2442-5G BB-2442-1G
COBBBB-2442-5GEA 0 EUR
COBBBB-2442-5G COBBBB-2442-1G
4-(Brommethyl)phenylboronsäure
4-(Brommethyl)phenylboronsäure
Formel: C₇H₈BBrO₂
MDL: MFCD01318113
CAS-Nummer: 68162-47-0
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR