4-Chlor-3-(trifluormethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3502-1G BB-3502-25G BB-3502-5G
COBBBB-3502-1GEA 0 EUR
COBBBB-3502-1G COBBBB-3502-25G COBBBB-3502-5G
4-Chlor-3-(trifluormethyl)phenylboronsäure
4-Chlor-3-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BClF₃O₂
MDL: MFCD03094999
CAS-Nummer: 176976-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR