4-Cyan-2-iodanilin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Amino-3-iodbenzoesäurenitril, 4-Amino-3-iodbenzonitril
H55400.03
H55400.06
H55400.03EA
35.9
EUR
H55400.03
H55400.06
4-Cyan-2-iodanilin 97%
4-Cyan-2-iodanilin
Purity: 97%
CAS No.: 33348-34-4
Molecular Formula: C7H5IN2
Formula Weight: 244.03
Melting Point: 112-115°
MDL No.: MFCD04039964
Formel:
C₇H₅IN₂ Molecular Weight: 244,03 g/mol Schmelzpunkt: 112…115 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039964 CAS-Nummer: 33348-34-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...