4-Amino-3-brombenzonitril 97%

Lieferant: Thermo Scientific
Gefahr

H55483.03 H55483.06
H55483.03EA 45.6 EUR
H55483.03 H55483.06
4-Amino-3-brombenzonitril 97%
4-Amino-3-brombenzonitril

Purity: 97% CAS No.: 50397-74-5 Molecular Formula: C7H5BrN2 Formula Weight: 197.03 Melting Point: 106-110° UN No.: 3439 MDL No.: MFCD01861386

Formel: C₇H₅BrN₂
Molecular Weight: 197,03 g/mol
Schmelzpunkt: 106…110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01861386
CAS-Nummer: 50397-74-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR