2-Amino-5-brombenzonitril 96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Bromo-2-cyanoaniline

H55217.03 H55217.06
H55217.03EA 44.7 EUR
H55217.03 H55217.06
2-Amino-5-brombenzonitril 96%
2-Amino-5-brombenzonitril

Purity: 96% CAS No.: 39263-32-6 Molecular Formula: C7H5BrN2 Formula Weight: 197.03 Melting Point: 96-100° UN No.: 3439 MDL No.: MFCD00158946

Formel: C₇H₅BrN₂
Molecular Weight: 197.03 g/mol
Schmelzpunkt: 96…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00158946
CAS-Nummer: 39263-32-6
EINECS: 254-387-8
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR