3-Brom-5-fluorphenol ≥97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-Brom-3-fluor-5-hydroxybenzol

H55288.03 H55288.06
H55288.03EA 20.2 EUR
H55288.03 H55288.06
3-Brom-5-fluorphenol ≥97%
3-Brom-5-fluorphenol

Purity: 97% CAS No.: 433939-27-6 Molecular Formula: C6H4BrFO Formula Weight: 191.00 Flash Point: 110°(230°F) Melting Point: 36-40° MDL No.: MFCD07783710

Formel: C₆H₄BrFO
Molecular Weight: 191 g/mol
Schmelzpunkt: 36…40 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD07783710
CAS-Nummer: 433939-27-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR