2-Chlor-4-hydroxybenzaldehyd ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H55771.03
H55771.06
H55771.03EA
171
EUR
H55771.03
H55771.06
2-Chlor-4-hydroxybenzaldehyd ≥97%
2-Chloro-4-hydroxybenzaldehyde
Purity: 97%
CAS No.: 56962-11-9
Molecular Formula: C7H5ClO2
Formula Weight: 156.57
Melting Point: 145-147°
MDL No.: MFCD00052184
Formel:
C₇H₅ClO₂ Molecular Weight: 156.57 g/mol Schmelzpunkt: 145…147 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052184 CAS-Nummer: 56962-11-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...