5-Cyan-3-pyridinboronsäurepinakolester 96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)nicotinonitrile, 3-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 3-Cyanopyridine-5-boronic acid Pinacol ester, 3-Cyanopyridine-5-boronic acid pinacol ester, 2-(5-Cyano-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

H54385.MD H54385.03
H54385.MDEA 56 EUR
H54385.MD H54385.03
5-Cyan-3-pyridinboronsäurepinakolester 96%
5-Cyan-3-pyridinboronsäurepinakolester

Purity: 96% CAS No.: 402718-29-0 Molecular Formula: C12H15BN2O2 Formula Weight: 230.07 Melting Point: 122-124° MDL No.: MFCD07780755

Formel: C₁₂H₁₅BN₂O₂
Molecular Weight: 230.07 g/mol
Schmelzpunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07780755
CAS-Nummer: 402718-29-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR