2-Cyan-5-pyridinboronsäurepinakolester 96%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
2-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile, 2-(6-Cyano-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-Cyano-3-pyridylboronic acid Pinacol ester, 2-Cyanopyridine-5-boronic acid pinacol ester
H54689.MD
H54689.03
H54689.MDEA
144
EUR
H54689.MD
H54689.03
2-Cyan-5-pyridinboronsäurepinakolester 96%
2-Cyan-5-pyridinboronsäurepinakolester
Purity: 96%
CAS No.: 741709-63-7
Molecular Formula: C12H15BN2O2
Formula Weight: 230.07
Melting Point: 105-106°
MDL No.: MFCD06657825
Formel:
C₁₂H₁₅BN₂O₂ Molecular Weight: 230.07 g/mol Schmelzpunkt: 105…106 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06657825 CAS-Nummer: 741709-63-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...