1-Phenyl-2-(trimethylsilyl)acetylen ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Trimethyl(phenylethynyl)silane
H55866.06
H55866.14
H55866.22
H55866.06EA
49.6
EUR
H55866.06
H55866.14
H55866.22
1-Phenyl-2-(trimethylsilyl)acetylen ≥99%
1-Phenyl-2-(trimethylsilyl)acetylen
Purity: 99%
CAS No.: 2170-06-1
Molecular Formula: C11H14Si
Formula Weight: 174.32
Flash Point: 74°(165°F)
Physical Form: Liquid
Density: 0.886
Refractive Index: 1.5280
Appearance: Colourless
Boiling Point: 87-88°/9mm
Beilstein Reference: 3088678
MDL No.: MFCD00054860
Formel:
C₁₁H₁₄Si Molecular Weight: 174,32 g/mol Siedepunkt: 87…88 °C (9 mmHg) Dichte: 0,886 Flammpunkt: 74 °C (165 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00054860 CAS-Nummer: 2170-06-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...