2,6-Di-tert-butylpyridin ≥97%

Lieferant: Thermo Scientific
Achtung

H55761.03 H55761.06 H55761.14
H55761.03EA 54.6 EUR
H55761.03 H55761.06 H55761.14
2,6-Di-tert-butylpyridin ≥97%
2,6-Di-tert-butylpyridin

Purity: 97+% CAS No.: 585-48-8 Molecular Formula: C13H21N Formula Weight: 191.32 Flash Point: 71°(160°F) Refractive Index: 1.4730 Boiling Point: 100-101°/23mm Beilstein Reference: 125886 Merck Reference: 14,3036 MDL No.: MFCD00006306

Formel: C₁₃H₂₁N
Molecular Weight: 191,32 g/mol
Siedepunkt: 100…101 °C (23 mmHg)
Dichte: 0,867
Flammpunkt: 71 °C (160 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006306
CAS-Nummer: 585-48-8
EINECS: 209-557-6
Merck Index: 14,3036

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR