(4-Fluorphenylthio)essigsäure

Lieferant: Thermo Scientific

Synonyme: 2-((4-Fluorophenyl)thio)acetic acid

KM09206DA KM09206EA
MAYBKM09206DAEA 0 EUR
MAYBKM09206DA MAYBKM09206EA
(4-Fluorphenylthio)essigsäure
(4-Fluorphenylthio)essigsäure
Formel: C₈H₇FO₂S
Molecular Weight: 186,21 g/mol
MDL: MFCD00219825
CAS-Nummer: 332-51-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR