(4-Bromphenyl)aceton ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 4-Bromophenylacetone

A19028.14 A19028.06
A19028.14EA 228 EUR
A19028.14 A19028.06
(4-Bromphenyl)aceton ≥98%
(4-Bromphenyl)aceton

Purity: 98% CAS No.: 6186-22-7 Molecular Formula: C9H9BrO Formula Weight: 213.07 MDL No.: MFCD00210401

Formel: C₉H₉BrO
Molecular Weight: 213.07 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00210401
CAS-Nummer: 6186-22-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR