Di-tert-butylchlorsilan
Lieferant: CHEMPUR SILANE
SILS05775.5
SILS05775.10
SILS05775.25
SILNSILS05775.5EA
0
EUR
SILNSILS05775.5
SILNSILS05775.10
SILNSILS05775.25
Di-tert-butylchlorsilan
Di-tert-butylchlorsilan
Formel:
C₈H₁₉ClSi Molecular Weight: 178.78 g/mol |
CAS-Nummer:
56310-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...