Di-tert-butyldichlorsilan, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
287660-25G
287660-5G
SIAL287660-25GEA
679
EUR
SIAL287660-25G
SIAL287660-5G
Di-tert-butyldichlorsilan, Sigma-Aldrich®
Di-tert-butyldichlorsilan
Formel:
C₈H₁₈Cl₂Si Molecular Weight: 213,22 g/mol |
MDL:
MFCD00008796 CAS-Nummer: 18395-90-9 EINECS: 242-273-0 UN: 2987 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...