Di-tert-butyldichlorsilan
Lieferant: CHEMPUR SILANE
SILS05825.5
SILS05825.25
SILNSILS05825.5EA
0
EUR
SILNSILS05825.5
SILNSILS05825.25
Di-tert-butyldichlorsilan
Di-tert-butyldichlorsilan
Formel:
C₈H₁₈Cl₂Si Molecular Weight: 213,22 g/mol |
MDL:
MFCD00008796 CAS-Nummer: 18395-90-9 EINECS: 242-273-0 UN: 2987 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...