Valeramid 97

Lieferant: Thermo Fisher Scientific

Synonyme: Pentanamide

L14601.06 L14601.14
L14601.06EA 33.6 EUR
L14601.06 L14601.14
Valeramid 97
Valeramid

Purity: 97% CAS No.: 626-97-1 Molecular Formula: CH3(CH2)3CONH2 Formula Weight: 101.15 Melting Point: 102-104° Beilstein Reference: 1740792

Formel: C₅H₁₀O₂
Molecular Weight: 102.13 g/mol
Schmelzpunkt: 102…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041895
CAS-Nummer: 626-97-1
EINECS: 210-974-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR