Kaliumhexachlororhodat(III) ≥99,99% (Metall-Basis) Rh ≥ 23.3%, Premion®

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Potassium hexachlororhodate(III)

41013.01 41013.03 41013.06
41013.01EA 152 EUR
41013.01 41013.03 41013.06
Kaliumhexachlororhodat(III) ≥99,99% (Metall-Basis) Rh ≥ 23.3%, Premion®
Potassium hexachlororhodate (III)

Purity: 99.99% CAS No.: 13845-07-3 Molecular Formula: K3RhCl6 Formula Weight: 432.93 Physical Form: Powder Sensitivity: Hygroscopic MDL No.: MFCD00192414

Formel: Cl₆K₃Rh
Molecular Weight: 432.92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00192414
CAS-Nummer: 13845-07-3
EINECS: 237-569-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR