n-Nonadecan-[D40] (98% D)

Lieferant: LGC Standards PROMOCHEM

Synonyme: n-Nonadecane-[²H₄₀ ]

CILDLM13460.1 CIL-DLM-1346-1
PROOCILDLM13460.1EA 155 EUR
PROOCILDLM13460.1 PROOCIL-DLM-1346-1
n-Nonadecan-[D40] (98% D)
n-Nonadecan-[D40]
Formel: C₁₉D₄₀
Molecular Weight: 308,21 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 39756-36-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR