Di-tert-butylsilan

Lieferant: FLUOROCHEM

019203-10G 019203-25G
FLUO019203-10GEA 0 EUR
FLUO019203-10G FLUO019203-25G
Di-tert-butylsilan
Di-tert-butylsilan
Formel: C₈H₂₀Si
Molecular Weight: 144,33 g/mol
MDL: MFCD00270996
CAS-Nummer: 30736-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR