Di-tert-butylsilan

Lieferant: CHEMPUR SILANE

SILS05880.5 SILS05880.25
SILNSILS05880.5EA 0 EUR
SILNSILS05880.5 SILNSILS05880.25
Di-tert-butylsilan
Di-tert-butylsilan
Formel: C₈H₂₀Si
Molecular Weight: 144,33 g/mol
MDL: MFCD00270996
CAS-Nummer: 30736-07-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR