4-Butoxyphenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(4-Butoxyphenyl)boronic acid, 4-n-Butoxyphenylboronic acid
H52865.03
H52865.06
H52865.03EA
33.4
EUR
H52865.03
H52865.06
4-Butoxyphenylboronsäure ≥98%
4-Butoxyphenylboronsäure
Purity: 98%
CAS No.: 105365-51-3
Molecular Formula: C10H15BO3
Formula Weight: 194.04
Melting Point: 102-104°
MDL No.: MFCD03427054
Formel:
C₁₀H₁₅BO₃ Molecular Weight: 194,04 g/mol Schmelzpunkt: 102…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03427054 CAS-Nummer: 105365-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...