2-Chlor-4-isopropylpyrimidin

Lieferant: COMBI-BLOCKS

HC-6113-5G HC-6113-1G
COBBHC-6113-5GEA 1070 EUR
COBBHC-6113-5G COBBHC-6113-1G
2-Chlor-4-isopropylpyrimidin
2-Chlor-4-isopropylpyrimidin
Formel: C₇H₉ClN₂
Molecular Weight: 156.61 g/mol
MDL: MFCD09475913
CAS-Nummer: 941294-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR