2-Methyl-3-indolylessigsäure ≥98%
Lieferant: Thermo Scientific
Synonyme:
2-Methylindole-3-acetic acid
L19071.03
L19071.06
L19071.03EA
31.2
EUR
L19071.03
L19071.06
2-Methyl-3-indolylessigsäure ≥98%
2-Methyl-3-indolylessigsäure
Purity: 98+%
CAS No.: 1912-43-2
Molecular Formula: C11H11NO2
Formula Weight: 189.22
Melting Point: 198-204° dec.
Beilstein Reference: 152701
Formel:
C₁₁H₁₁NO₂ Molecular Weight: 189,21 g/mol Schmelzpunkt: 198…204 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075006 CAS-Nummer: 1912-43-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...