2-Butoxy-5-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8143-1G BB-8143-250M
COBBBB-8143-1GEA 0 EUR
COBBBB-8143-1G COBBBB-8143-250M
2-Butoxy-5-fluorphenylboronsäure
2-Butoxy-5-fluorphenylboronsäure
Formel: C₁₀H₁₄BFO₃
MDL: MFCD05664232
CAS-Nummer: 480438-62-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR