2-Butoxy-6-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-8174-1G BB-8174-250M
COBBBB-8174-1GEA 88.4 EUR
COBBBB-8174-1G COBBBB-8174-250M
2-Butoxy-6-fluorphenylboronsäure
2-Butoxy-6-fluorphenylboronsäure
Formel: C₁₀H₁₄BFO₃
Molecular Weight: 212,03 g/mol
MDL: MFCD06798079
CAS-Nummer: 870777-19-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR