3-Butoxy-2,6-difluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(3-Butoxy-2,6-difluorophenyl)boronic acid
BB-8202-1G
BB-8202-250M
COBBBB-8202-1GEA
143
EUR
COBBBB-8202-1G
COBBBB-8202-250M
3-Butoxy-2,6-difluorphenylboronsäure
3-Butoxy-2,6-difluorphenylboronsäure
Formel:
C₁₀H₁₃BF₂O₃ Molecular Weight: 230,02 g/mol |
MDL:
MFCD07369754 CAS-Nummer: 849062-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...