Dibutylamin 1% in DMF

Lieferant: Bernd Kraft
Gefahr

Synonyme: N-butyl-1-propanamin

14752.2700 14752.3700 14752.4700
KRAF14752.2700EA 0 EUR
KRAF14752.2700 KRAF14752.3700 KRAF14752.4700
Dibutylamin 1% in DMF
Dibutylamin
Formel: C₈H₁₉N
Molecular Weight: 129,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00009429
CAS-Nummer: 111-92-2
EINECS: 203-921-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0d7d86589224f8565e12dc8e6b30a8de923393f&vid=d2cc023db25785e54555fc105aa8436e65fd9cbc&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR