Dibutylamin 1 mol/l (1 N) in Toluol

Lieferant: Bernd Kraft

Synonyme: N-butyl-1-propanamin

10462.3700 10462.2700 10462.4700
KRAF10462.3700EA 134 EUR
KRAF10462.3700 KRAF10462.2700 KRAF10462.4700
Dibutylamin 1 mol/l (1 N) in Toluol
Dibutylamin
Formel: C₈H₁₉N
Molecular Weight: 129,25 g/mol
MDL: MFCD00009429
CAS-Nummer: 111-92-2
EINECS: 203-921-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0d7d86589224f8565e12dc8e6b30a8de923393f&vid=d2cc023db25785e54555fc105aa8436e65fd9cbc&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR