Chloroform : isoamyl alcohol (24:1) (v/v)

Lieferant: Uptima
Achtung

UP899254 UP899255
UPTIUP899254EA 0 EUR
UPTIUP899254 UPTIUP899255
Chloroform : isoamyl alcohol (24:1) (v/v)
Chloroform : isoamyl alcohol
Formel: CHCl₃,C₅H₁₂O
Lagertemperatur: Raumtemperatur
UN: 1888
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR