3-Methylbutylisovalerat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Isovaleric acid Isopentyl ester, Isovaleric acid Isoamyl ester, Isoamyl Isovalerate, Isopentyl 3-methylbutanoate, Isopentyl Isovalerate
M-Clarity™ Qualitätsstufe = MQ300
W208507-1KG-K
W208507-20KG-K
W208507-9KG-K
SIALW208507-1KG-KEA
80.6
EUR
SIALW208507-1KG-K
SIALW208507-20KG-K
SIALW208507-9KG-K
3-Methylbutylisovalerat, Sigma-Aldrich®
3-Methylbutylisovalerat
Formel:
C₁₀H₂₀O₂ Molecular Weight: 172,27 g/mol |
MDL:
MFCD00048369 CAS-Nummer: 659-70-1 EINECS: 211-536-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...