Hexamethyldisilathian ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: Bis(trimethylsilyl)sulphide

39556.03 39556.06
39556.03EA 55.1 EUR
39556.03 39556.06
Hexamethyldisilathian ≥98%
Hexamethyldisilathian

Purity: 98% CAS No.: 3385-94-2 Molecular Formula: [(CH3)3Si]2S Formula Weight: 178.44 Flash Point: 26°(79°F) Physical Form: Liquid in ampoules Density: 0.851 Refractive Index: 1.4598 Odour: Pungent Boiling Point: 162-166° Beilstein Reference: 1740046 UN No.: 1993 MDL No.: MFCD00014851

Formel: C₆H₁₈SSi₂
Molecular Weight: 178,45 g/mol
Siedepunkt: 162…166 °C
Dichte: 0,851
Flammpunkt: 26 °C (79 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014851
CAS-Nummer: 3385-94-2
EINECS: 222-201-4
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR