Hexamethyldisilathian

Lieferant: CHEMPUR SILANE

SILS02800.1 SILS02800.5
SILNSILS02800.1EA 0 EUR
SILNSILS02800.1 SILNSILS02800.5
Hexamethyldisilathian
Hexamethyldisilathian
Formel: C₆H₁₈SSi₂
MDL: MFCD00014851
CAS-Nummer: 3385-94-2
EINECS: 222-201-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR