Bis(4-fluorphenyl)disulfid ≥98%

Lieferant: Thermo Scientific
Achtung

H32589.03 H32589.06
H32589.03EA 52.3 EUR
H32589.03 H32589.06
Bis(4-fluorphenyl)disulfid ≥98%
Bis(4-fluorphenyl)disulfid

Purity: 98% CAS No.: 405-31-2 Molecular Formula: C12H8F2S2 Formula Weight: 254.31 Boiling Point: 141°/1mm MDL No.: MFCD00816758

Formel: C₁₂H₈F₂S₂
Molecular Weight: 254,32 g/mol
Siedepunkt: 141 °C (1 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00816758
CAS-Nummer: 405-31-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR