Gold(III)hydroxid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Gold(iii)hydroxide
M-Clarity™ Qualitätsstufe = MQ100
398462-1G
398462-5G
SIAL398462-1GEA
220
EUR
SIAL398462-1G
SIAL398462-5G
Gold(III)hydroxid, Sigma-Aldrich®
Gold trihydroxide
Formel:
H₃AuO₃ Molecular Weight: 247.99 |
MDL:
MFCD00046173 CAS-Nummer: 1303-52-2 EINECS: 215-120-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...