(S)-(-)-3-Chlor-1-phenyl-1-propanol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(S)-3-Chloro-1-phenylpropan-1-ol
324612-1G
324612-5G
SIAL324612-1GEA
31.2
EUR
SIAL324612-1G
SIAL324612-5G
(S)-(-)-3-Chlor-1-phenyl-1-propanol, Sigma-Aldrich®
(S)-(-)-3-Chlor-1-phenyl-1-propanol
Formel:
C₉H₁₁ClO Molecular Weight: 170.64 g/mol |
MDL:
MFCD00013309 CAS-Nummer: 100306-34-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...