(2-Chlorphenyl)aceton 96%

Lieferant: Thermo Scientific

Synonyme: 1-(2-Chlorophenyl)propan-2-one

L09846.03 L09846.06
L09846.03EA 23.4 EUR
L09846.03 L09846.06
(2-Chlorphenyl)aceton 96%
(2-Chlorphenyl)aceton

Purity: 96% CAS No.: 6305-95-9 Molecular Formula: C9H9ClO Formula Weight: 168.62 Refractive Index: 1.5360 Boiling Point: 102-103°/3mm MDL No.: MFCD00045175

Formel: C₉H₉ClO
Molecular Weight: 168,62 g/mol
Siedepunkt: 102…103 °C (3 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00045175
CAS-Nummer: 6305-95-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR