2-Chlor-4-phenylpyrimidin

Lieferant: COMBI-BLOCKS

HC-6171-1G HC-6171-5G
COBBHC-6171-1GEA 153 EUR
COBBHC-6171-1G COBBHC-6171-5G
2-Chlor-4-phenylpyrimidin
2-Chloro-4-phenylpyrimidine
Formel: C₁₀H₇ClN₂
Molecular Weight: 190.63 g/mol
MDL: MFCD00234600
CAS-Nummer: 13036-50-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR