Cadmiumacetat 4%, reinst

Lieferant: Bernd Kraft

03026.6020 03026.2000 03026.3000 03026.4000
KRAF03026.6020EA 398 EUR
KRAF03026.6020 KRAF03026.2000 KRAF03026.3000 KRAF03026.4000
Cadmiumacetat 4%, reinst
Cadmiumacetat
Formel: (H₃CCOO)₂Cd
Molecular Weight: 230.5 g/mol
Schmelzpunkt: 256
Dichte: 2.34
MDL: MFCD00012449
CAS-Nummer: 543-90-8
EINECS: 208-853-2
Merck Index: 12,01650

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR