Cadmiumacetat 5%, reinst

Lieferant: Bernd Kraft

03120.6020 03120.2000 03120.3000 03120.4000
KRAF03120.6020EA 398 EUR
KRAF03120.6020 KRAF03120.2000 KRAF03120.3000 KRAF03120.4000
Cadmiumacetat 5%, reinst
Cadmiumacetat
Formel: (H₃CCOO)₂Cd
MDL: MFCD00012449
CAS-Nummer: 543-90-8
EINECS: 208-853-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR